Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM7 Antibody (S74), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP212906Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212906Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212906Z Supplier Novus Biologicals Supplier No. NBP212906AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TRPM7 Monoclonal specifically detects TRPM7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TRPM7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S74-25
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK
Gene Symbols TRPM7
Host Species Mouse
Immunogen Fusion protein amino acids 1817-1863 (C- terminus) of mouse TrpM7
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 54822
Test Specificity Detects approx 220kDa. No cross-reactivity against TrpM6.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.