Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPM7, p Ser1493 Antibody, Novus Biologicals™
SDP

Catalog No. NB387868 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387868 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387868 Supplier Novus Biologicals Supplier No. NBP323179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPM7 Polyclonal antibody specifically detects TRPM7 in Human samples. It is validated for Western Blot

Specifications

Antigen TRPM7
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias CHAK, CHAK1FLJ25718, Channel-kinase 1, EC 2.7.11.1, Long transient receptor potential channel 7, LTRPC ion channel family member 7, LTrpC7, LTrpC-7, LTRPC7FLJ20117, transient receptor potential cation channel subfamily M member 7, transient receptor potential cation channel, subfamily M, member 7, transient receptor potential-phospholipase C-interacting kinase, TRP-PLIK
Host Species Rabbit
Immunogen Phospho-TRPM7 (Ser-1493) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding Ser-1493 in human TRPM7. This site is conserved in rat and mouse TRPM7, but the homology surrounding the site is low. This site has no homology to other TRPM family members. [UniProt# Q96QT4]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 54822
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.