Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRPV3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA511471
Change view
Click to view available options
Quantity:
400 μL
Catalog No. Quantity
PIPA511471 400 μL
1 options

Catalog No. PIPA511471

Supplier: Invitrogen™ PA511471

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPV3 Polyclonal Antibody for Western Blot, IHC (P)

TRPV3 belongs to a family of nonselective cation channels that function in a variety of processes, including temperature sensation and vasoregulation. The thermosensitive members of this family are expressed in subsets of sensory neurons that terminate in the skin, and are activated at distinct physiological temperatures. This channel is activated at temperatures between 22 and 40 degrees C. This gene lies in close proximity to another family member (TRPV1) gene on chromosome 17, and the two encoded proteins are thought to associate with each other to form heteromeric channels.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRPV3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TRPV3
Gene Accession No. Q8K424
Gene Alias 1110036I10Rik; AI644701; FNEPPK2; heat sensitive channel TRPV3; MGC124324; MGC124325; Nh; non-hair; OLMS; Transient receptor potential cation channel subfamily V member 3; transient receptor potential cation channel, subfamily V, member 3; TRP ion channel TRPV3; TrpV3; Vanilloid receptor-like 3; vanilloid receptor-related osmotically activated channel protein; VRL 3; VRL3; VRL-3
Gene Symbols TRPV3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 90-119 amino acids from the N-terminal region of mouse TRPV3.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246788
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.