Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRPV4 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB343573 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343573 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343573 Supplier Novus Biologicals Supplier No. NBP241262MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRPV4 Polyclonal antibody specifically detects TRPV4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TRPV4
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CMT2Cosmosensitive transient receptor potential channel 4, HMSN2C, Osm-9-like TRP channel 4, OTRPC4SSQTL1, transient receptor potential cation channel subfamily V member 4, transient receptor potential cation channel, subfamily V, member 4, Transient receptor potential protein 12, TRP12OSM9-like transient receptor potential channel 4, TrpV4, Vanilloid receptor-like channel 2, Vanilloid receptor-like protein 2, vanilloid receptor-related osmotically activated channel, Vanilloid receptor-related osmotically-activated channel, VRL2, VRL-2, VR-OAC, VROACSPSMA
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid peptide near the center of human TRPV4. The immunogen is located within amino acids 380 - 430 of TRPV4. Amino Acid Squence: RREVTDEDTRHLSRKFKD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 59341
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.