Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrueCut™ Cas9 Protein v2

Catalog No. p-8370003
Encompass_Preferred
Click to view available options
Quantity:
10 μg
100 μg
25 μg
500 μg
Concentration:
1 μg/μL
5 μg/μL

A next-generation CRISPR Cas9 protein engineered to deliver maximum on-target editing efficiency. TrueCut Cas9 Protein v2 is ideal for applications requiring the highest editing efficiency.

Invitrogen TrueCut Cas9 Protein v2 is a next-generation CRISPR Cas9 protein engineered to deliver maximum editing efficiency.

Features include:

  • Consistently high editing efficiency in all tested cell lines including standard, immune, primary, and stem, with up to 2X higher editing efficiency in difficult targets than the competition
  • High quality—manufactured under strict ISO 13485 quality standards
  • Validated protocols for a large number of cell types help you achieve success faster

TrueCut Cas9 Protein v2 is recombinant Streptococcus pyogenes Cas9 (wt) protein, purified from E. coli, for genome editing with CRISPR technology. Cas9 protein forms a very stable ribonucleoprotein (RNP) complex with the guide RNA (gRNA) component of the CRISPR/Cas9 system. Incorporation of nuclear localization signals (NLS) aids delivery to the nucleus, increasing the rate of genomic DNA cleavage.

  • Transfection-ready, using either lipid-mediated transfection reagents or electroporation
  • Eliminates time-consuming cloning steps
  • Available in two concentrations:
    • 1 μg/μL for use in standard editing scenarios
    • 5 μg/μL for optimization of editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time
    • For custom sizes or concentrations, please inquire

Options for obtaining CRISPR gRNA for use with TrueCut Cas9 Protein v2:

  • Order pre-designed TrueGuide Synthetic gRNAs for high-efficiency knock out in human or mouse
  • Submit your own design for custom synthesis of CRISPR sgRNA
  • Use our free TrueDesign CRISPR Genome Editor software for easy design and ordering of custom gRNA for precise genome editing

Certifications

CoA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 μg/μL
Components TrueCut Cas9 Protein v2
Grade Molecular Biology
Content And Storage TrueCut Cas9 Protein v2 (5 mg/mL), 100 μg

Store at -5°C to -30°C.
Accession Number NC_002737.2:854751-858857
Expression System E. coli
Gene ID (Entrez) 901176
Gene Symbol Spy_1046
Gene Alias cas5, csn1
Protein Form Recombinant
Protein Tag Untagged
Shipping Condition Dry Ice
Validated Application Genome Editing
Quantity 100 μg
Product Type Cas9 Protein v2
Research Category Genome Editing
Product Line TrueCut™
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.