Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrueGuide™ sgRNA Positive Control, Rosa26 (mouse)

Catalog No. a35525
Encompass_Preferred
Click to view available options
Quantity:
3 nmol

Catalog No. A35525

Supplier: Invitrogen™ A35525

Only null left
Add to Cart
Edge
Add to Cart

Used for the optimization of transfection and verification of editing efficiency in transfected cells.

Provides complete collection of validated positive controls for the TrueGuide Synthetic gRNA product line to help maximize editing performance. This positive control against ROSA26 targets intronic region of gene and has achieved editing efficiencies of up to 80% in variety of mouse cell lines. ROSA26 control easily determines the optimal conditions for delivering CRISPR/Cas9 tools to cell type, allowing to move forward in editing experiments and minimize downstream effort to isolate and validate edited clones

Validated positive control crRNA targeting Rosa26

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Delivered as dried RNA. Store dried RNA at room temperature; store resuspended RNA at -5°C to -30°C. Resuspend in nuclease-free water or TE.
Format Tube
Control Type Positive Control
RNAi Type gRNA
Species Mouse
Product Line TrueGuide
Product Type gRNA
Quantity 3 nmol
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.