Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRUSS Antibody, Novus Biologicals™
SDP

Catalog No. NBP247593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247593 0.1 mL
NB396156 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247593 Supplier Novus Biologicals Supplier No. NBP247593
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TRUSS Polyclonal specifically detects TRUSS in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TRUSS
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias C20orf188, dJ756N5.2, DKFZp586C1223, DKFZP727M231, Protein TAP1, Protein TRUSS, short transient receptor potential channel 4 associated protein, short transient receptor potential channel 4-associated protein, TNF-receptor ubiquitous scaffolding/signaling protein, transient receptor potential cation channel, subfamily C, member 4 associatedprotein, Trp4-associated protein, Trpc4-associated protein, TRRP4APchromosome 20 open reading frame 188, TRUSS, tumor necrosis factor receptor-associated ubiquitous scaffolding and signalingprotein
Gene Symbols TRPC4AP
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: LIWRKHSASALVLHGHNQNCDCSPDITLKIQFLRLLQSFSDHHENKYLLLNNQELNELSAISLKANIPEVEAVLNTDRSLVCDGK
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26133
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.