Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin 1/PRSS1 Antibody, Novus Biologicals™
SDP

Catalog No. NB6001277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001277 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001277 Supplier Novus Biologicals Supplier No. NB6001277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trypsin 1/PRSS1 Polyclonal specifically detects Trypsin 1/PRSS1 in Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Trypsin 1/PRSS1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:5000-1:25000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P00760
Gene Alias Beta-trypsin, Cationic trypsinogen, digestive zymogen, DKFZp779A0837, EC 3.4.21, EC 3.4.21.4, FLJ57296, MGC120175, MGC149362, nonfunctional trypsin 1, protease, serine, 1 (trypsin 1), Serine protease 1, TRP1, TRY1beta-trypsin, TRY4, TRYP1, Trypsin I, trypsin-1, trypsinogen 1, trypsinogen A
Gene Symbols PRSS1
Host Species Rabbit
Immunogen Trypsin 1/PRSS1 1/PRSS1 [Bovine Pancreas] (Uniprot: P00760)
Molecular Weight of Antigen 25.79 kDa
Purification Method Delipidation and Defibrination
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5644
Test Specificity Trypsin
Target Species Bovine
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.