Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin 3/PRSS3 Antibody (001), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB068894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB068894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB068894 Supplier Novus Biologicals Supplier No. NBP290010IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Trypsin 3/PRSS3 Monoclonal specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for ELISA.

Specifications

Antigen Trypsin 3/PRSS3
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 755
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin III, Trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Trypsin 3/PRSS3 (P35030-3; Met1-Ser247).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 5646
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.