Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin 3/PRSS3 Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB323244 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323244 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323244 Supplier Novus Biologicals Supplier No. NBP299895JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trypsin 3/PRSS3 Polyclonal antibody specifically detects Trypsin 3/PRSS3 in Human samples. It is validated for ELISA

Specifications

Antigen Trypsin 3/PRSS3
Applications ELISA
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Brain trypsinogen, EC 3.4.21, EC 3.4.21.4, Mesotrypsinogen, pancreatic trypsinogen III, protease, serine, 3, protease, serine, 3 (mesotrypsin), protease, serine, 4 (trypsin 4, brain), PRSS4, Serine protease 3, Serine protease 4, T9, TRY3MTG, TRY4mesotrypsin, trypsin 3, Trypsin III, Trypsin IV, trypsin-3, trypsinogen 4, trypsinogen 5, trypsinogen IV
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Trypsin 3/PRSS3 (Accession#: P35030-3; Met 1-Ser 247)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 5646
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.