Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Trypsin-EDTA (0.5%), no phenol red
GREENER_CHOICE

Catalog No. 15400054
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-400-054 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-400-054 Supplier Gibco™ Supplier No. 15400054
Only null left
Add to Cart
Add to Cart

Made from trypsin powder, an irradiated mixture of proteases derived from porcine pancreas

Due to its digestive strength, trypsin solution is widely used for cell dissociation, routine cell culture passaging, and primary tissue dissociation. Concentrated forms of trypsin can be aseptically diluted to 1X using a balanced salt solution without calcium and magnesium. The trypsin concentration required for dissociation varies with cell type and experimental requirements.

  • Solutions tested for pH, osmolality, sterility, and performance
  • Prior to manufacturing, the raw materials are verified for e-beam irradiation and tested for endotoxin, PPV, PCV 1/2, mycoplasma, bacterial, fungal, and viral contamination, as well as multiple activity assays, ash analysis, and moisture analysis
  • Documented Traceability available including lot traceability, animal origin certificates, lot analyses, irradiation certificates, a viral inactivation summary, and supply chain transparency

Sustainable packaging

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Compliance

Manufactured at a cGMP compliant facility that is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Chelators EDTA
Content And Storage Storage conditions: -5°C to -20°C
Cell Type Mammalian Cells
Classification Animal Origin
Concentration 10 X
Form Liquid
Product Type Cell Culture Dissociation Reagent
Sterility Sterile-filtered
With Additives EDTA
Without Additives No Phenol Red
Osmolality 280 - 340 mOsm/kg
pH 7.1 to 8.0
Product Line Gibco
Quantity 100 mL
Shipping Condition Wet Ice
Source Animal Origin
Tested For In vitro Bioassay
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.