Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Trypsin from Beef pancreas

Catalog No. MP021011795 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021011795 500 mg
MP210117901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP021011795 Supplier MP Biomedicals™ Supplier No. 02101179.5
Add to Cart
Add to Cart

Used to dissolve blood clots in its microbial form and treat inflammation in its pancreatic form.

Applications: Trypsin can be used to dissolve blood clots in its microbial form and treat inflammation in its pancreatic form. Trypsin is commonly used in biological research.
Product Description: Trypsin consists of a single chain polypeptide of 223 amino acid residues. Trypsin is a member of the serine protease family.
Biochem/physiol Actions: Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.
Unit Definition: One unit hydrolyzes one µmole of TAME per minute at 25 °C, pH 8.2, in the presence of ionized calcium.
Typical Working Concentration: For the Removal of Adherent Cells from a Culture Surface: 2.5% w/v solution
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH Optimum pH: Approximately 8.0 (Lit.)
For Use With (Application) Dissolve blood clots
Content And Storage 4°C
Quantity 500 mg
Product Type Proteins, Enzymes & Peptides
Molecular Weight (g/mol) Trypsin is composed of two subunits, α-trypsin and β-trypsin. α-Trypsin is composed of two peptide chains and β-trypsin is composed of one chain.
Form Lyophilized Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.