Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Trypsin from Beef pancreas
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mg
1 g
Description
Applications: Trypsin can be used to dissolve blood clots in its microbial form and treat inflammation in its pancreatic form. Trypsin is commonly used in biological research.
Product Description: Trypsin consists of a single chain polypeptide of 223 amino acid residues. Trypsin is a member of the serine protease family.
Biochem/physiol Actions: Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.
Unit Definition: One unit hydrolyzes one µmole of TAME per minute at 25 °C, pH 8.2, in the presence of ionized calcium.
Typical Working Concentration: For the Removal of Adherent Cells from a Culture Surface: 2.5% w/v solution
Product Description: Trypsin consists of a single chain polypeptide of 223 amino acid residues. Trypsin is a member of the serine protease family.
Biochem/physiol Actions: Trypsin cleaves peptides on the C-terminal side of lysine and arginine residues. The rate of hydrolysis of this reaction is slowed if an acidic residue is on either side of the cleavage site and hydrolysis is stopped if a proline residue is on the carboxyl side of the cleavage site. Trypsin can also act to cleave ester and amide linkages of synthetic derivatives of amino acids. EDTA is added to trypsin solutions as a chelating agent that neutralizes calcium and magnesium ions that obscure the peptide bonds on which trypsin acts. Removing these ions increases the enzymatic activity.
Unit Definition: One unit hydrolyzes one µmole of TAME per minute at 25 °C, pH 8.2, in the presence of ionized calcium.
Typical Working Concentration: For the Removal of Adherent Cells from a Culture Surface: 2.5% w/v solution
Specifications
Specifications
| Color | White |
| pH | Optimum pH: Approximately 8.0 (Lit.) |
| For Use With (Application) | Dissolve blood clots |
| Content And Storage | 4°C |
| Quantity | 500 mg |
| Product Type | Proteins, Enzymes & Peptides |
| Molecular Weight (g/mol) | Trypsin is composed of two subunits, α-trypsin and β-trypsin. α-Trypsin is composed of two peptide chains and β-trypsin is composed of one chain. |
| Form | Lyophilized Powder |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction