Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin Inhibitor Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NBP17773801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17773801 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17773801 Supplier Novus Biologicals Supplier No. NBP1777380.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trypsin Inhibitor Polyclonal specifically detects Trypsin Inhibitor in Plant samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Trypsin Inhibitor
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot 1:500 - 1:3000, ELISA 1:5000 - 1:25000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Gene Alias KTI3, Kunitz trypsin inhibitor
Gene Symbols KTI3
Host Species Rabbit
Immunogen Trypsin Inhibitor [Lima Bean]
Purification Method Delipidation and Defibrination
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 547831
Target Species Plant
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.