Learn More
Description
Specifications
Specifications
| Antigen | Trypsin Inhibitor |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:5000, ELISA 1:5000-1:20000, Immunoprecipitation 1:100 |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | P01070 |
| Gene Alias | KTI3, Kunitz trypsin inhibitor |
| Gene Symbols | KTI3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
