Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Trypsin Inhibitor, chicken egg whites

Catalog No. p-7082382
Click to view available options
Quantity:
500 mg
1 g
2 g

Inhibits the pancreatic enzyme trypsin | CAS: 9035-81-8

Soybean trypsin inhibior (STI) is a protein which reduces trypsin (a proteolytic enzyme) as well as plasma kallikrein, factor Xa and plasmin activity. Soybean trypsin inhibitor has demonstrated inhibition of human neutrophil elastase at an IC50=0.3 nM. At this concentration soybean trypsin inhibitor showed neither cytotoxic nor haemolytic effects on populations of human blood cells. In fibrin matrix, and without toxic effects, STI had a strong inhibitory effect on human umbilical vein endothelial cells migration and tubulogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9035-81-8
Synonym Trypsin inhibitor, ovomucoid

Specifications

Quantity 500 mg
Description White
For Use With (Application) Inhibits the pancreatic enzyme trypsin
Product Type Signal Transduction Reagent
Form Powder
EINECSNumber 232-906-9
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.