Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsinogen Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NBP17769750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17769750 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17769750 Supplier Novus Biologicals Supplier No. NBP17769750UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Trypsinogen Polyclonal specifically detects Trypsinogen in Bovine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Trypsinogen
Applications ELISA, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:1000 - 1:5000, ELISA 1:10000 - 1:40000
Formulation 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, 10Mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate
Gene Alias 2, Brain trypsinogen, Cationic Trypsinogen, Mesotrypsinogen, Pancreatic anionic trypsinogen, Protease, PRSS2, PRSS3, PRSS4, serine, TRP 1, TRP1, TRY1, TRY2, TRY3. TRY4, TRYP1, TRYP2, TRYP8, Trypsin 2, Trypsin 3, Trypsin III, Trypsin IV
Host Species Rabbit
Immunogen Trypsinogen [Bovine Pancreas]. Uniprot ID Q29463
Purification Method Delipidation and Defibrination
Quantity 50 μg
Primary or Secondary Primary
Test Specificity Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum as well as purified and partially purified Trypsinogen from bovine pancreas.
Reconstitution Reconstitute with 100μL deionized water.
Target Species Bovine
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.