Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/beta 1 Antibody (AA1), Novus Biologicals™
SDP

Catalog No. NB019404 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019404 0.05 mL
NBP226444 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019404 Supplier Novus Biologicals Supplier No. NBP2264440.05ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tryptase alpha/TPS1 Monoclonal specifically detects Tryptase alpha/TPS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Tryptase alpha/TPS1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunofluorescence, Immunocytochemistry, ELISA, Flow Cytometry
Classification Monoclonal
Clone AA1
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:200, Immunohistochemistry-Frozen 1:10-1:500
Formulation Citrate, pH 6.0, containing less than 0.1% sodium azide with Sodium Azide
Gene Alias TPS1, TPS2, TPSAB1, TPSB1, tryptase alpha/beta 1
Gene Symbols TPSAB1
Host Species Mouse
Immunogen Purified human mast cell tryptase was used as immunogen (Walls et al, 1990).
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7177
Test Specificity Rat and mouse reactivity reported in scientific literature. Please see Publications tab for full list.
Target Species Human, Rat, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.