Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/beta 1 Antibody (AA1), Novus Biologicals™
SDP

Catalog No. NBP226444 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP226444 0.25 mL
NB019404 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP226444 Supplier Novus Biologicals Supplier No. NBP226444
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen Tryptase alpha/beta 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AA1
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:300-1:600, Immunohistochemistry-Paraffin 1:300-1:600, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Alias EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III
Gene Symbols TPSAB1
Host Species Mouse
Immunogen BALB/C mice were injected with a purified human lung tryptase.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Immunology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7177
Test Specificity Due to sequence identity this antibody will likely cross react with tryptase beta 2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.