Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/beta 1 Antibody (RM0355-10H34), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP211983UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211983UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211983UV Supplier Novus Biologicals Supplier No. NBP211983UV
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Tryptase alpha/beta 1
Applications Western Blot
Classification Monoclonal
Clone RM0355-10H34
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q02844
Gene Alias EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III
Gene Symbols TPSAB1
Host Species Rat
Immunogen Mouse recombinant Tryptase alpha/beta 1
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Immunology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7177
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.