Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase alpha/beta 1 Antibody (RM0355-10H34), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB361513 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361513 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361513 Supplier Novus Biologicals Supplier No. NBP211983MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Tryptase alpha/beta 1 Monoclonal antibody specifically detects Tryptase alpha/beta 1 in Mouse samples. It is validated for Western Blot

Specifications

Antigen Tryptase alpha/beta 1
Applications Western Blot
Classification Monoclonal
Clone RM0355-10H34
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC:3.4.21.59, epididymis secretory sperm binding protein, mast cell alpha II tryptase, mast cell beta I tryptase, mast cell tryptase, TPS1, TPS2, TPSB1, TPSB2, tryptase alpha II, Tryptase alpha-1, tryptase beta I, tryptase beta-1, Tryptase beta-2, Tryptase I, tryptase II, tryptase-1, tryptase-2, tryptase-I, tryptase-III
Host Species Rat
Immunogen Mouse recombinant Tryptase alpha/beta 1
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7177
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.