Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase beta-2/TPSB2 Antibody (03), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB353208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB353208 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB353208 Supplier Novus Biologicals Supplier No. NBP306181MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tryptase beta-2/TPSB2 Monoclonal antibody specifically detects Tryptase beta-2/TPSB2 in Human samples. It is validated for ELISA

Specifications

Antigen Tryptase beta-2/TPSB2
Applications ELISA
Classification Monoclonal
Clone 03
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2tryptase beta-2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Tryptase beta-2/TPSB2 (Accession#: NP_077078.5; Met 1-Pro 275).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 64499
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.