Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase beta-2/TPSB2 Antibody (9H6), Novus Biologicals™
SDP

Catalog No. NB298431 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298431 100 μL
NB298430 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298431 Supplier Novus Biologicals Supplier No. NBP326273100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tryptase beta-2/TPSB2 Monoclonal antibody specifically detects Tryptase beta-2/TPSB2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen Tryptase beta-2/TPSB2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 9H6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias EC 3.4.21, EC 3.4.21.59, mast cell tryptase beta II, mast cell tryptase beta III, TPS2tryptase beta-2, tryptase beta 2, tryptase beta 2 (gene/pseudogene), tryptase beta II, tryptase beta III, Tryptase II, tryptase III, tryptase-2, tryptaseB, tryptaseC
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Tryptase beta-2/TPSB2 [UniProt P20231]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 64499
Target Species Human, Mouse
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.