Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase gamma-1/TPSG1 Antibody (OTI1E11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274649A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274649A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274649A700 Supplier Novus Biologicals Supplier No. NBP274649AF700

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Tryptase gamma-1/TPSG1 Monoclonal specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen Tryptase gamma-1/TPSG1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1E11
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II
Gene Symbols TPSG1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 20-283 of human Tryptase gamma-1/TPSG1 (NP_036599) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 25823.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.