Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptase gamma-1/TPSG1 Antibody (OTI1E11), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB282276 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB282276 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB282276 Supplier Novus Biologicals Supplier No. NBP274649MFV450

Mouse Monoclonal Antibody

Tryptase gamma-1/TPSG1 Monoclonal antibody specifically detects Tryptase gamma-1/TPSG1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Tryptase gamma-1/TPSG1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1E11
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.21, EC 3.4.21.-, gamma I, gamma II, lung tryptase, mast cell protease II, mast cell tryptase, PRSS31skin tryptase, Serine protease 31, TMTpituitary tryptase, Transmembrane tryptase, trpA, tryptase gamma, tryptase gamma 1, tryptase gamma I, tryptase gamma II
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 20-283 of human TPSG1(NP_036599) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25823
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.