Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ Trypticase Soy Broth (TSB) Bottled Broth 100mL

Catalog No. 10486161 Shop All Osmonics Products
Click to view available options
Broth Type:
TSB Double Strength Broth
TSB Single Strength Broth
Formulation:
Double Strength
Single Strength

Trypticase Soy Broth (TSB), Bottled Broth is a medium that will support the growth of a wide variety of Microorganisms including aerobic, facultative, and anaerobic bacteria and fungi.

Description

  • General purpose medium used in qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms. Trypticase Soy Broth – Single Strength complies with the demands of the DIN Norm 10167 for the detection of Escherichia coli serotype 0157:H7 in foods and FDA-BAM for the isolation of enterohemorrhagic Escherichia coli (EHEC). In addition the media conforms to the formula of the US Pharmacopoeia.

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 100 mL bottled solution, Reduce Risk of Cross-Contamination.
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • General purpose medium used in qualitative procedures for the cultivation of fastidious and non fastidious organisms.
  • TSB is a medium that will support the growth of a wide variety of microorganisms including aerobic, facultative, and anaerobic bacteria and fungi.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits.
This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Broth Type TSB Double Strength Broth
Certifications/Compliance Batch-specific quality certificate in each pack
Detects cultivation of fasticious and non-fastidious microorganisms.
Sterility Sterile
Formulation Double Strength
No. per Pack 1
Packaging Type Bottle
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.