Learn More
Tryptophan Hydroxylase 1/TPH-1 Antibody (145), DyLight 755, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Tryptophan Hydroxylase 1/TPH-1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 145 |
| Conjugate | DyLight 755 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Tryptophan Hydroxylase 1/TPH-1 (P17752-1; Ile2-Ile444). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.