Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tryptophan Hydroxylase 1/TPH-1 Antibody (145), FITC, Novus Biologicals™
SDP

Catalog No. NB334852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334852 Supplier Novus Biologicals Supplier No. NBP290022F
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tryptophan Hydroxylase 1/TPH-1 Monoclonal antibody specifically detects Tryptophan Hydroxylase 1/TPH-1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Tryptophan Hydroxylase 1/TPH-1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 145
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 1.14.16, EC 1.14.16.4, indoleacetic acid-5-hydroxylase, L-tryptophan hydroxylase, MGC119994, TPH, TPRH, TRPH, tryptophan 5-hydroxylase 1, Tryptophan 5-monooxygenase 1, tryptophan hydroxylase (tryptophan 5-monooxygenase), tryptophan hydroxylase 1
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Tryptophan Hydroxylase 1/TPH-1 (Uniprot#: P17752-1, Ile2-Ile444)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 7166
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.