Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSBP Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB170184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170184 25 μg
NB170183 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB170184 Supplier Novus Biologicals Supplier No. NBP31755425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSBP Polyclonal antibody specifically detects TSBP in Human samples. It is validated for Immunofluorescence

Specifications

Antigen TSBP
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias C6orf10, chromosome 6 open reading frame 10, testis specific basic protein, TSBP
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ILTGYMDEELAKKSCSKIQILKCGGTARSQNSREENKEALKNDIIFTNSVESLKSAHIKEPEREGKGTDLEKDKIGMEVKVDSDAGIP
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 10665
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.