Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176618SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176618SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176618SS Supplier Novus Biologicals Supplier No. NBP176618SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSC1 Polyclonal specifically detects TSC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TSC1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AAC51674
Gene Alias KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor
Gene Symbols TSC1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from the middle region of human TSC1. The immunogen is located within amino acids 220 - 270 of TSC1. Amino Acid Squence: KDHELDPRRWKRLE
Molecular Weight of Antigen 135 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Primary or Secondary Primary
Gene ID (Entrez) 7248
Test Specificity At least two isoforms of TSC1 are known to exist; this antibody will detect both isoforms. TSC1 antibody is predicted to not cross-react with TSC2.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.