Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176619 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176619 Supplier Novus Biologicals Supplier No. NBP176619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSC2 Polyclonal specifically detects TSC2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.

Specifications

Antigen TSC2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4 ug/ml, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_000539
Gene Alias LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein
Gene Symbols TSC2
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the amino terminus of human TSC2. The immunogen is located within the first 50 amino acids of TSC2. Amino Acid Squence: TPRPNPRSAEGKQT
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7249
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.