Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC22/TSC22D1 Antibody (043), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB037650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB037650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB037650 Supplier Novus Biologicals Supplier No. NBP290143AF647
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TSC22/TSC22D1 Monoclonal specifically detects TSC22/TSC22D1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TSC22/TSC22D1
Applications Immunofluorescence
Classification Monoclonal
Clone 43
Conjugate Alexa Fluor 647
Dilution Immunocytochemistry/Immunofluorescence, Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TSC22/TSC22D1 (Q15714-2; Met1-Ala144).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8848
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.