Learn More
Description
Specifications
Specifications
| Antigen | TSC22/TSC22D1 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 143 |
| Conjugate | Janelia Fluor 646 |
| Dilution | Immunocytochemistry/Immunofluorescence, Immunofluorescence |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human TSC22/TSC22D1 (Q15714-2; Met1-Ala144). |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
