Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSEN34 Antibody, Novus Biologicals™
SDP

Catalog No. NBP181315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP181315 0.1 mL
NB419063 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP181315 Supplier Novus Biologicals Supplier No. NBP181315
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSEN34 Polyclonal specifically detects TSEN34 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TSEN34
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 3.1.27.9, hsSen34, leukocyte receptor cluster (LRC) member 5, Leukocyte receptor cluster member 5, PCH2C, SEN34LENG5SEN34L, tRNA splicing endonuclease 34 homolog (S. cerevisiae), tRNA splicing endonuclease 34 homolog (SEN34, S. cerevisiae), tRNA-intron endonuclease Sen34, tRNA-splicing endonuclease subunit Sen34
Gene Symbols TSEN34
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:WERGFFLSAAGKFGGDFLVYPGDPLRFHAHYIAQCWAPEDTIPLQDLVAAGRLGTSVRKTLLLCSPQPDGKVVYTSLQW
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79042
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.