Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSG Antibody (RM0140-7D19), PerCP, Novus Biologicals™
SDP

Catalog No. NB361962 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361962 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361962 Supplier Novus Biologicals Supplier No. NBP122516PCP
Add to Cart
Add to Cart

Rat Monoclonal Antibody

TSG Monoclonal antibody specifically detects TSG in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen TSG
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RM0140-7D19
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias TSGtwisted gastrulation protein homolog 1, twisted gastrulation homolog 1 (Drosophila)
Host Species Rat
Immunogen Mouse recombinant TSG
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57045
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.