Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSG101 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB336814 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336814 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336814 Supplier Novus Biologicals Supplier No. NBP277452APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TSG101 Polyclonal antibody specifically detects TSG101 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen TSG101
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TSG101 protein (between amino acids 125-210) [UniProt Q99816]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Microautophagy
Primary or Secondary Primary
Gene ID (Entrez) 7251
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.