Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSH beta Antibody (154) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10062385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062385 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062385 Supplier Novus Biologicals Supplier No. NB10062385
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSH beta Monoclonal specifically detects TSH beta in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TSH beta
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 154
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Accession No. P01222
Gene Alias thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA
Gene Symbols TSHB
Host Species Mouse
Immunogen Native Human beta-TSH
Purification Method Protein A or G purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7252
Test Specificity This product is specific for is specific for beta thyroid stimulating hormone (beta TSH), a hormone produced by the pituitary gland, which acts to stimulate the growth of the thyroid and production of thyroid hormones. This antibody shows minimal cross-reactivity with beta Human chorionic gonadotropin (hCG), Luteinizing Hormone (LH) or Follicle Stimulating Hormone (FSH).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.