Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSH beta Antibody (TSH-116), Biotin, Novus Biologicals™
SDP

Catalog No. NB500363B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500363B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500363B Supplier Novus Biologicals Supplier No. NB500363B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSH beta Monoclonal specifically detects TSH beta in Human samples. It is validated for ELISA.

Specifications

Antigen Thyroid Stimulating Hormone
Applications ELISA
Classification Monoclonal
Clone TSH-116
Conjugate Biotin
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA
Gene Symbols TSHB
Host Species Mouse
Immunogen Human thyrotropin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7252
Test Specificity This antibody (clone TSH-116) reacts with human thyroid stimulating hormone (hTSH, thyrotropin), a glycoprotein hormone produced by the anterior pituitary gland cells in response to signals from the hypothalamus gland in the brain. The TSH-116 reacts with association constant 1.1 x 1011 l/mol. Following cross-reactivity expressed as binding of labelled hormone (% of total) was determined by solid phase RIA with excess of the antibody TSH-116: hTSH (78.9), hCG (20.3), hLH (23.2), hFSH (29.9).
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.