Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHR Antibody (484404), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB176370 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176370 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176370 Supplier Novus Biologicals Supplier No. FAB65342Z
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen TSHR
Applications Flow Cytometry
Classification Monoclonal
Clone 484404
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TSH R, Accession # P16473
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7253
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.