Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHR Antibody (484404), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346952 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346952 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346952 Supplier Novus Biologicals Supplier No. FAB65342MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSHR Monoclonal antibody specifically detects TSHR in Human samples. It is validated for Flow Cytometry

Specifications

Antigen TSHR
Applications Flow Cytometry
Classification Monoclonal
Clone 484404
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human TSH R, Accession # P16473
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7253
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.