Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHR Antibody (4C1), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. N065797AP01 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N065797AP01 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N065797AP01 Supplier Novus Biologicals Supplier No. NB10065797APC0.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen TSHR
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4C1
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% sodium azide
Gene Alias CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Gene Symbols TSHR
Host Species Mouse
Immunogen External domain of human TSH receptor.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7253
Test Specificity Recognizes the human thyroid stimulating hormone (TSH) receptor (otherwise known as the thyrotropin receptor) binding to an extracellular domain.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.