Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHR Antibody (TSHRB/1404), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP254395G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254395G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP254395G Supplier Novus Biologicals Supplier No. NBP254395G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen TSHR
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TSHRB/1404
Conjugate DyLight 488
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Gene Symbols TSHR
Host Species Mouse
Immunogen Recombinant human TSHR protein (Uniprot: P16473)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7253.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.