Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSHZ2 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB184013 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184013 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184013 Supplier Novus Biologicals Supplier No. NBP282041R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSHZ2 Polyclonal antibody specifically detects TSHZ2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen TSHZ2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias C20orf17, cell growth-inhibiting protein 7, DKFZp686K2480, teashirt homolog 2, teashirt zinc finger homeobox 2, ZABC2, zinc finger protein 218
Host Species Rabbit
Immunogen TSHZ2 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human TSHZ2. The immunogen is located within amino acids 30 - 80 of TSHZ2. Amino Acid Squence: NDTGTDEELETGPEQKG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128553
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.