Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Boronate-5PW HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021830 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
5 mm
7.5 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
11-102-1830 7.5 mm
11-102-1850 5 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-1830 Supplier MilliporeSigma Supelco Supplier No. 813066
Add to Cart
Add to Cart

High efficiency, resin-based affinity columns separate or purify many enzymes and other proteins.

  • High efficiency, resin-based affinity columns separate or purify many enzymes and other proteins
  • Coupling of m-aminophenyl boronate to TSKgel 5PW polymeric support results in a ligand capable of forming a tetrahedral boronate anion under alkaline pH conditions
  • This anionic structure can bind with 1,2 cis-diol groups such as those found in carbohydrates, carbohydrate containing compounds, and catecholamines
  • Interaction between the boronate anion and the 1,2 cis-diol groups is enhanced in the presence of Mg2+ ions and is inhibited by amine-containing buffers
  • Adsorption onto the matrix takes place in basic buffers such as HEPES and morpholine
  • Desorption takes place in carbohydrate or amine-containing mobile phases such as sorbitol or Tris
  • Resin Adsorption Capacity: 40 μmol/mL
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 1.2 mL/min
Length (Metric) 7.5 cm
Max. Pressure 30 bar
Particle Size 10 μm
pH 2 to 9
Pore Size 1000 Å
Solvents ≤ 20% Organic Solvent
Temperature >0°C
Stationary Phase M-Aminophenyl Boronate
Product Line TSKgel
Sample Type Proteins
Matrix M-Aminophenyl Boronate Phase/Hydroxylated Methacrylic Polymer
Technique Affinity
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.