Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Ether-5PW HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021814 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric):
2 mm
5 mm
7.5 mm
8 mm
Material:
Glass
Stainless Steel
Stainless Steel Hardware
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
11-102-1814 7.5 mm Stainless Steel
11-102-1844 5 mm Glass
11-102-1845 8 mm Glass
11-102-1973 2 mm Stainless Steel Hardware
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-1814 Supplier MilliporeSigma Supelco Supplier No. 808641
Add to Cart
Add to Cart

An excellent choice for separating hydrophobic molecules such as membrane proteins, or monoclonal antibodies such as IgG or IgM.

  • Among the TSKgel HIC Columns, TSKgel Ether-5PW has intermediate hydrophobity
  • An excellent choice for separating hydrophobic molecules such as membrane proteins or monoclonal antibodies such as IgG or IgM
  • Features a 1000 Å pore size in a polymethacrylate base material
  • Column is stable in either acid or caustic cleaning regimens
  • Provides excellent access to larger molecules with low diffusion coefficients
  • Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity allowing for selective binding, and desorption of proteins
  • Recommended for purifying more hydrophobic proteins
  • Salt Concentration: ≤4 M
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.5 mm
Flow Rate 1.2 mL/min
Length (Metric) 7.5 cm
Max. Pressure 20 bar
Particle Size 10 μm
pH 2 to 12
Pore Size 1000 Å
Solvents ≤ 50% Organic Solvent
Temperature 10°C to 50°C
Stationary Phase Ether
Product Line TSKgel
Shipping Condition Shipped aq. soln.
Matrix Ether Phase/Hydroxylated Methacrylic Polymer
Material Stainless Steel
Technique HIC HPLC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.