Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ HHR Column, 13 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021903 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1903 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1903 Supplier MilliporeSigma Supelco Supplier No. 817393

TSKgel™ GMHhr-M Mixed Bed Columns are packed with rigid, multiple pore size, polymer beads. Mixed bed columns are commonly used for unknown samples, and for the analysis of polymers with a broad molecular weight distribution.

  • Conventional GPC columns with dimensions of 7.8 mm ID x 30 cm
  • TSKgel GMHhr-M(S) 13 μm columns are recommended to prevent or limit shearing of ultra high molecular weight polymers
  • All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a broad range of organic solvents
  • Maximum Operating Temperature: 140°C
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7.8 mm
Flow Rate 0.5 to 1.0 mL/min
Length (Metric) 30 cm
Max. Pressure 20 bar
Particle Size 13 μm
Temperature 80°C
USP Type L21
Stationary Phase Polystyrene-divinylbenzene
Product Line TSKgel
Shipping Condition Shipped in THF
Material Stainless Steel
Technique SEC
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.