Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKgel™ Super-Phenyl HPLC Column, 2.3 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022037 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
5 cm
10 cm
Diameter (Metric):
2 mm
4.6 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-2037 5 cm 2 mm
11-102-2038 10 cm 2 mm
11-102-1937 5 cm 4.6 mm
11-102-1938 10 cm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-102-2037 Supplier MilliporeSigma Supelco Supplier No. 820017
Add to Cart
Add to Cart

TSKgel™ Super-Phenyl Columns are packed with monodispersed 2 μm (nominal, mean particle size is 2.3 μm) spherical silica particles covalently bonded with phenyl groups.

  • The small particle size used in Super Series and TSKgel ODS-140HTP Columns provide the highest efficiency of all TSKgel reversed phase columns
  • Monodispersity of the particles reduces column back pressure to values more typical of larger particles, allowing the use of higher flow rates compared to other 2 μm packings
  • TSKgel Super-Phenyl Columns are less hydrophobic than TSKgel Super-Octyl and TSKgel Super-ODS Columns
  • An excellent choice for peptides, proteins, amino acids, tryptic digests, nucleotides, pharmaceutical molecules and food/beverage samples
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 2 mm
Flow Rate 0.25 mL/min
Length (Metric) 5 cm
Max. Pressure 80 bar
Particle Shape Sphere
Particle Size 2.3 μm
pH 2 to 7.5
Solvents 0-100% Organic Solvent
Surface Area 100 m2/g
Temperature 10°C to 50°C
Stationary Phase Phenyl
Product Line TSKgel
Shipping Condition Shipped in 50% Acetonitrile, 50% Water
Matrix Phenyl Phase/Silica Particle Platform
Carbon Load 3%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.