missing translation for 'onlineSavingsMsg'
Learn More

TSKgel™ TMS-250 HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™

Catalog No. 111021774 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
7.5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
11-102-1774 7.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-1774 Supplier MilliporeSigma Supelco Supplier No. 807190

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Featuring trimethylsilane groups covalently attached to large 250 Å pores, TSKgel™ TMS-250 is an excellent choice for analysis of larger proteins by reversed phase HPLC.

Due to the low hydrophobicity of the ligand, excellent recoveries are common even when used with large proteins. Proteins such as adolase (158 kDa) exhibit sharp peaks relative to wide pore C18 columns.

Order Info

Shipped in 50% methanol - 50% water.

TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 7.5 cm
Max. Pressure 200 bar
Particle Size 10 μm
pH 2.0 - 7.5
Pore Size 250 Å
Solvents 0 to 100% organic solvent
Temperature 10°C to 80°C
USP Type L1, L23
Stationary Phase C1 (methyl)
Product Line TSKgel
Matrix Silica Particle Platform
Carbon Load 5%
Endcapped Yes
Phase Reversed Phase
Material Stainless Steel Hardware
Show More Show Less

TSKgel is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.