Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSKS Antibody, Novus Biologicals™
SDP

Catalog No. NBP220749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220749 Supplier Novus Biologicals Supplier No. NBP220749

Rabbit Polyclonal Antibody

TSKS Polyclonal specifically detects TSKS in Human samples. It is validated for Western Blot.

Specifications

Antigen TSKS
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UJT2
Gene Alias STK22 substrate 1, STK22S1, testis specific serine/threonine kinase substrate, Testis-specific kinase substrate, testis-specific serine kinase substrate, TSKS1, TSSKS
Gene Symbols TSKS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TSKS. The exact sequence is proprietary.
Molecular Weight of Antigen 65 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 60385
Test Specificity Human. Species reactivity based on sequence identity: Mouse (90%), Rat (89%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.