Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSLP Antibody (55N1D10), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP227326FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227326FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227326FR Supplier Novus Biologicals Supplier No. NBP227326FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TSLP Monoclonal specifically detects TSLP in Human, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TSLP
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 55N1D10
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry-Paraffin
Gene Accession No. Q969D9
Gene Alias thymic stromal lymphopoietin
Gene Symbols TSLP
Host Species Mouse
Immunogen A partial recombinant TSLP protein (amino acids 27-158) was used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85480
Target Species Human, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.